missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromic Acid, 10% w/v, Reagents
Specifications
Specifications
| Color | Orange |
| Boiling Point | 101°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.071 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Percent Purity | 10% w/v |
| Grade | Reagent |
| Chemical Name or Material | Chromic Acid, 10% w/v |