missing translation for 'onlineSavingsMsg'
Learn More

Chromic Acid, 10% (w/v), Ricca Chemical

Catalog No. 207716
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
207716 500 mL Amber Glass Bottle
207732 1 L Amber Glass Bottle
20771 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 207716 Supplier Ricca Chemical Company Supplier No. 207716

Chemical Identifiers

CAS 7732-18-5 , 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Color Orange
Concentration 10% (w/v)
CAS Min % 89.0, 9.0
CAS Max % 93.0, 11.0
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Percent Purity 10% W/V
Grade Laboratory
Chemical Name or Material Chromic Acid
Show More Show Less