Learn More
Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 320
Super thick grade is one of the thickest papers on the market. It offers extremely high flow rate and is recommended both for electrophoresis of large molecules and as a blotter to complete the blotting sandwich in gel transfer applications.
Supplier:  Ahlstrom-Munksjo 32082020
 View more versions of this product
Catalog No. 09-790-54K
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
High
320
2.45 mm
200 x 200 mm
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
Fine, Coarse
Western Blot Filter Paper
Product Suggestions
Product Suggestions