missing translation for 'onlineSavingsMsg'
Learn More

Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 320

Catalog No. 0979054G
Change view
Click to view available options
Dimensions (L x W):
100 x 150 mm
150 x 150 mm
200 x 200 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
0979054G 150 x 150 mm
0979054D 100 x 150 mm
0979054K 200 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0979054G Supplier Ahlstrom-Munksjo Supplier No. 32081515
Only null left

Super thick grade is one of the thickest papers on the market. It offers extremely high flow rate and is recommended both for electrophoresis of large molecules and as a blotter to complete the blotting sandwich in gel transfer applications.

Specifications

Flow Rate High
For Use With (Application) Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
Grade 320
Thickness (Metric) 2.45 mm
Dimensions (L x W) 150 x 150 mm