missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150011
Description
CHRNE Polyclonal Antibody for Western Blot, ICC/IF, IHC
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.Specifications
| CHRNE | |
| Polyclonal | |
| Unconjugated | |
| CHRNE | |
| acetylcholine receptor; acetylcholine receptor epsilon; acetylcholine receptor subunit epsilon; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; Achre; AChrepsilon; ACHRG; Acre; cholinergic receptor; cholinergic receptor nicotinic epsilon subunit; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; CHRNE; CMS1D; CMS1E; CMS2A; CMS4A; CMS4B; CMS4C; epsilon-subunit; FCCMS; gene for acetylcholine receptor epsilon subunit; MGC133376; nAChRE; nicotinic acetylcholine receptor epsilon subunit; SCCMS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11448, 1145, 29422 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P09660, P20782, Q04844 | |
| CHRNE | |
| Recombinant Cholinergic Receptor, Nicotinic, Epsilon (CHRNe) (Lys21-Lys239). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |