missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRNA5 Recombinant Rabbit Monoclonal Antibody (23GB3775)
GREENER_CHOICE

Catalog No. MA552578
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552578 100 μL
MA552579 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552578 Supplier Invitrogen™ Supplier No. MA552578
Only null left

Rabbit Recombinant Monoclonal Antibody

CHRNA5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.

Specifications

Antigen CHRNA5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3775
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHRNA5
Gene Accession No. P20420, P30532, Q2MKA5
Gene Alias acetylcholine receptor; acetylcholine receptor alpha 5; acetylcholine receptor, nicotinic, alpha 5 (neuronal); Acra5; Acra-5; cholinergic receptor; cholinergic receptor nicotinic alpha 5 subunit; Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5; cholinergic receptor, nicotinic alpha 5; cholinergic receptor, nicotinic, alpha 5; cholinergic receptor, nicotinic, alpha 5 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 5; CHRNA5; hypothetical protein LOC550584; LNCR2; nAChR alpha 5; NACHRA5; Neuronal acetylcholine receptor subunit alpha-5; neuronal nicotinic acetylcholine receptor alpha5 subunit; neuronal nicotinic acetylcholine receptor, alpha5 subunit; nicotinic acetylcholine receptor alpha5 subunit; zgc:110642
Gene Symbols CHRNA5
Host Species Rabbit
Immunogen A synthetic peptide of human CHRNA5 (200-250AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110835, 1138, 25102
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less