missing translation for 'onlineSavingsMsg'
Learn More

Chondroitin Sulfate Sodium Salt Dietary Supplement, USP, Spectrum™ Chemical

C1610, 9082-07-9

$2757.10 - $31278.40

Chemical Identifiers

CAS 9082-7-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601683
View Documents
Spectrum Chemical Mfg Cor
C1610500GM
500 g Poly Bottle
Each for $2,757.10
Only null left
 
18601684
View Documents
Spectrum Chemical Mfg Cor
C161025KG
2.5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18601685
View Documents
Spectrum Chemical Mfg Cor
C161025KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Chondroitin Sulfate Sodium Salt, USP Dietary Supplement is the sodium salt of a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid) and is used for osteoarthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

9082-7-9

Specifications

9082-7-9
500 g
20%-30%
Poly Bottle
100%
USP Dietary Supplement
12%
-20.0° to -30.0°
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'