missing translation for 'onlineSavingsMsg'
Learn More

Chondroitin Sulfate Sodium Salt Dietary Supplement, USP, Spectrum™ Chemical

Catalog No. 18601683
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601683 500 g Poly Bottle
18601684 2.5 kg Poly Pail
18601685 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601683 Supplier Spectrum Chemical Mfg Cor Supplier No. C1610500GM
Only null left

C1610, 9082-07-9

Spectrum™ Chemical Chondroitin Sulfate Sodium Salt, USP Dietary Supplement is the sodium salt of a sulfated glycosaminoglycan (GAG) composed of a chain of alternating sugars (N-acetylgalactosamine and glucuronic acid) and is used for osteoarthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9082-7-9

Specifications

CAS Max % 100%
Quantity 500 g
Grade USP Dietary Supplement
Ignition Residue 20%-30%
Loss on Drying 12%
Packaging Poly Bottle
Specific Rotation -20.0° to -30.0°