missing translation for 'onlineSavingsMsg'
Learn More

Choline Dihydrogen Citrate 99.0+%, TCI America™

$52.19 - $203.17

Chemical Identifiers

CAS 77-91-8
Molecular Formula C11H21NO8
MDL Number MFCD00038734
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C033125G
View Documents
TCI America
C033125G
25 g N/A
Only null left
 
C0331500G
View Documents
TCI America
C0331500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77-91-8
C11H21NO8
MFCD00038734

Specifications

77-91-8
White
MFCD00038734
295.29
Crystalline Powder
107°C
C11H21NO8
25 g
≥99.0% (T)
Choline Dihydrogen Citrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GA2582000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'