missing translation for 'onlineSavingsMsg'
Learn More

Choline Dihydrogen Citrate 99.0+%, TCI America™

Catalog No. C0331500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0331500G 500 g
C033125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0331500G Supplier TCI America Supplier No. C0331500G
Only null left

Chemical Identifiers

CAS 77-91-8
Molecular Formula C11H21NO8
MDL Number MFCD00038734

Specifications

Melting Point 107°C
Color White
Quantity 500 g
Formula Weight 295.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Choline Dihydrogen Citrate
RTECSNumber GA2582000
TSCA Yes