missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Dihydrogen Citrate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 107°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 295.29 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Choline Dihydrogen Citrate |
Safety and Handling
| RTECSNumber | GA2582000 |
| TSCA | Yes |