missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015134
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015134 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015134 Supplier Merck Emd Millipore Supplier No. PHR1251500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 67-48-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (CH3)3 N(Cl)CH2 CH2 OH
MDL Number MFCD00011721
Melting Point 302°C to 305°C
Synonym (2-Hydroxyethyl)trimethylammonium chloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 139.62
Grade Certified Reference Material
Show More Show Less