missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cholic Acid, Sodium Salt, Calbiochem™,
Chemical Identifiers
| CAS | 361-09-1 |
|---|---|
| Molecular Formula | C24H39NaO5 |
| Molecular Weight (g/mol) | 430.56 |
| MDL Number | MFCD00150749,MFCD00064138 |
| InChI Key | NRHMKIHPTBHXPF-TUJRSCDTSA-M |
| PubChem CID | 57349315 |
| IUPAC Name | sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| SMILES | [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
22910150GM
|
MilliporeSigma™
22910150GM |
50 g | Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
| 361-09-1 | |
| 430.56 | |
| NRHMKIHPTBHXPF-TUJRSCDTSA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| C24H39NaO5 | |
| MFCD00150749,MFCD00064138 | |
| 57349315 | |
| [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
Specifications
| White | |
| C24H39NaO5 | |
| NRHMKIHPTBHXPF-TUJRSCDTSA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate | |
| 57349315 |
| 361-09-1 | |
| MFCD00150749,MFCD00064138 | |
| [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C | |
| 430.56 | |
| >96% (TLC) |