missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl benzoate, 98%

Catalog No. p-3757304
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC110201000 100 g Glass bottle
1 options

Catalog No. AC110201000

Supplier: Thermo Scientific Chemicals 110201000

Only null left

CAS: 604-32-0 | C34H50O2 | 490.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 604-32-0
Molecular Formula C34H50O2
Molecular Weight (g/mol) 490.77
MDL Number MFCD00003635
InChI Key UVZUFUGNHDDLRQ-UHFFFAOYNA-N
Synonym cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
PubChem CID 2723613
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1

Specifications

Melting Point 149°C to 150°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 10, 2181
Specific Rotation -15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,methylene chloride,,benzene,toluene,diethyl ether
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] benzoate
Formula Weight 490.76
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Cholesteryl benzoate
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-064-3

RUO – Research Use Only