missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, ≥99%, MP Biomedicals™

$146.49 - $146.49

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00003646
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10138205
View Documents
MP Biomedicals Inc
0210138205
5 g
Each for $146.49
Only null left
 
Description

Description

Specifications

Chemical Identifiers

57-88-5
386.664
HVYWMOMLDIMFJA-DPAQBDIFSA-N
5997
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
C27H46O
MFCD00003646
cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
CHEBI:16113
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

57-88-5
1.067g/mL
C27H46O
5 g
HVYWMOMLDIMFJA-DPAQBDIFSA-N
(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
5997
386.64
Cholesterol
White
≥99%
MFCD00003646
cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C
386.664
CHEBI:16113
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-353-2

Recommended Storage : Store at -0°C.

SDS
missing translation for 'documents'

missing translation for 'documents'