missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CHO-S-SFM II, with hypoxanthine and thymidine
GREENER_CHOICE

Catalog No. 12052098
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12052098 1 L
12052114 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12052098 Supplier Gibco™ Supplier No. 12052098

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHO-S-SFM II, with hypoxanthine and thymidine

CHO-S-SFM II, is a complete, serum-free, low-protein (less than 100μ/mL) medium optimized for the growth and maintenance of Chinese hamster ovary (CHO) cells in suspension culture for production of recombinant proteins. It is supplemented with hypoxanthine and thymidine (the non-supplemented equivalent is also available). This ready-to-use medium is suitable for the suspension culturing of CHO and other cells in batch, continuous, and perfusion culture systems. A CHO-S-SFM II medium Drug Master File (DMF) has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.

This medium contains components of animal origin. If growth factors or hydrolysates, found in competitor media, are to be avoided, consider CD CHO, CD OptiCHO™ or CD FortiCHO™ media.

When to use CHO-S-SFM II Medium
  • Your cells have been successfully cultured in CHO-S-SFM II medium in the past and your process is nearing a regulatory filing (DMF available)

    When to Consider Another Gibco™ Medium

    Consider CD FortiCHO™ medium when:
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    Consider CD OptiCHO™ medium when:
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    Consider CD CHO medium when:
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    Consider CD DG44 medium when:
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Specifications

    Cell Line CHO
    Classification Low Protein, Serum-free
    Concentration 1 X
    Form Liquid
    Product Type Serum Free Medium (SFM)
    Serum Level Serum-free
    With Additives Glutamine, Sodium Pyruvate, Sodium Bicarbonate, Thymidine, Hypoxanthine
    Without Additives No Phenol Red
    Product Line Gibco
    Quantity 1 L
    Shipping Condition Room Temperature
    Endotoxin Level Low
    Format Bottle(s)
    Included Antibiotics No antibiotics
    Content And Storage Store in refrigerator (2–8°C) and protect from light.
    Culture Type Suspension Cell Culture
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.