missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CHO CD EfficientFeed™ B AGT™ Nutrient Supplement
Description
Gibco™ CHO CD EfficientFeed™ B AGT™ nutrient supplement is a single part dry format product for fed batch addition to the culture of multiple cell lines. CHO CD EfficientFeed™ B AGT™ (Advanced Granulation Technology™) nutrient supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.
The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid format for small scale use and experimentation.
Expect up to 2- to 5-fold increases in productivity
Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
Simplify scale-up by leveraging the value of AGT™ simple reconstitution
Achieve performance comparable to our original liquid catalog product
Ready for larger scale use with convenient packaging options (10 L and 100 L)
Which EfficientFeed™ Nutrient Supplement to Choose?
Consider the CHO CD Efficient Feed™ Kit when:
You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0
Consider CHO CD EfficientFeed™ A supplement when:
You are currently using CD OptiCHO™ as your base cell growth medium
Consider CHO CD EfficientFeed™ B supplement when:
You are currently using CD CHO as your base cell growth medium
Consider CD EfficientFeed™ C AGT™ supplement when:
You want higher productivity in GS CHO and other selected cell lines
You are currently using CD FortiCHO™ as your base cell growth medium
You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0
Increased Titers
CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including hydrolysates (Figures 2, 3a, & 3b).
Simple Scale-Up and Cost Savings
The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ B AGT™ nutrient supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L catalog liquid as a seamless process, as well as allow for cost savings when purchasing production level volumes.
Comparable Performance to Liquid Supplement
Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ B (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.
AGT™ Format Ease of Use
CHO CD EfficientFeed™ B AGT™ nutrient supplement delivers dry format scale-up convenience as well as simple reconstitution via water addition and a single pH adjustment step (see Product Profile Sheet below for complete instructions).
For research or further manufacturing use. Not for use in diagnostic procedures.
The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid format for small scale use and experimentation.
Which EfficientFeed™ Nutrient Supplement to Choose?
Consider the CHO CD Efficient Feed™ Kit when:
Consider CHO CD EfficientFeed™ A supplement when:
Consider CHO CD EfficientFeed™ B supplement when:
Consider CD EfficientFeed™ C AGT™ supplement when:
Increased Titers
CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including hydrolysates (Figures 2, 3a, & 3b).
Simple Scale-Up and Cost Savings
The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ B AGT™ nutrient supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L catalog liquid as a seamless process, as well as allow for cost savings when purchasing production level volumes.
Comparable Performance to Liquid Supplement
Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ B (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.
AGT™ Format Ease of Use
CHO CD EfficientFeed™ B AGT™ nutrient supplement delivers dry format scale-up convenience as well as simple reconstitution via water addition and a single pH adjustment step (see Product Profile Sheet below for complete instructions).
For research or further manufacturing use. Not for use in diagnostic procedures.
Specifications
Specifications
| Content And Storage | Dry format medium Store at 2 to 8°C, Store Dark and Dry |
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Culture Type | Suspension Cell Culture |
| Form | AGT |
| Product Type | Nutrient Supplement |
| Serum Level | Serum-Free |
| Sterility | Non-sterile |
| With Additives | Sodium Pyruvate, Sodium Bicarbonate |
| Without Additives | No Glutamine, No Phenol Red, No Insulin |
| Show More |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.