missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHML Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101802
Description
Antibody detects endogenous levels of total CHML.
Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.Specifications
| CHML | |
| Polyclonal | |
| Unconjugated | |
| CHML | |
| AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1122, 12663 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26374, Q9QZD5 | |
| CHML | |
| A synthesized peptide derived from human CHML(Accession P26374), corresponding to amino acid residues V127-E177. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |