missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHM Recombinant Rabbit Monoclonal Antibody (23GB3065)
GREENER_CHOICE

Catalog No. MA552540
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552540 100 μL
MA552541 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552540 Supplier Invitrogen™ Supplier No. MA552540
Only null left

Rabbit Recombinant Monoclonal Antibody

CHM Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Substrate-binding subunit of the Rab geranylgeranyltransferase (GGTase) complex. Binds unprenylated Rab proteins and presents the substrate peptide to the catalytic component B composed of RABGGTA and RABGGTB, and remains bound to it after the geranylgeranyl transfer reaction. The component A is thought to be regenerated by transferring its prenylated Rab back to the donor membrane. Besides, a pre-formed complex consisting of CHM and the Rab GGTase dimer (RGGT or component B) can bind to and prenylate Rab proteins; this alternative pathway is proposed to be the predominant pathway for Rab protein geranylgeranylation.

Specifications

Antigen CHM
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3065
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHM
Gene Accession No. P24386, P37727, Q9QXG2
Gene Alias CHM; CHM, Rab escort protein 1; choroideraemia protein homolog; choroideremia (Rab escort protein 1); Choroideremia protein; Choroideremia protein homolog; choroidermia; choroidermia (RAB escort protein 1); DXS540; GGTA; HSD-32; Rab escort protein 1; rab proteins geranylgeranyltransferase component A 1; REP1; REP-1; TCD; TCD protein
Gene Symbols CHM
Host Species Rabbit
Immunogen A synthesized peptide derived from CHM (49-81AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1121, 12662, 24942
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less