missing translation for 'onlineSavingsMsg'
Learn More

Chlorzoxazone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015959
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015959 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015959 Supplier Merck Emd Millipore Supplier No. PHR1919500MG
Only null left

Specifications

CAS 95-25-0
Linear Formula C7H4 ClNO2
MDL Number MFCD00005717
Melting Point 191°C to 192°C
Synonym Chlorzoxazone; 5-Chloro-2(3 H)-benzoxazolone; 5-Chloro-2-hydroxybenzoxazole
Type Certified Reference Material
Quantity 500 mg
Formula Weight 169.57
Grade Certified Reference Material