missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone, 98%

Catalog No. AC460932500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460932500 250 mg
AC460930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460932500 Supplier Thermo Scientific Chemicals Supplier No. 460932500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-36-1 | C14H11ClN2O4S | 338.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-36-1
Molecular Formula C14H11ClN2O4S
Molecular Weight (g/mol) 338.77
InChI Key JIVPVXMEBJLZRO-UHFFFAOYSA-N
Synonym chlorthalidone, chlortalidone, phthalamudine, chlorphthalidolone, hygroton, chlorthalidon, phthalamodine, chlorothalidone, chlorphthalidone, natriuran
PubChem CID 2732
ChEBI CHEBI:3654
IUPAC Name 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
SMILES C1=CC=C2C(=C1)C(=O)NC2(C3=CC(=C(C=C3)Cl)S(=O)(=O)N)O

Specifications

Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 338.77
Percent Purity 98%
Chemical Name or Material Chlorthalidone
EINECSNumber 201-022-5

RUO – Research Use Only