missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012230
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012230 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012230 Supplier Merck Emd Millipore Supplier No. PHR1520500MG

missing translation for 'validMainframeMsg'

Chlortetracycline Hydrochloride is a salt of chlortetracycline, which is a tetracycline which shows broad-spectrum antibiotic activity. It is generally used as a veterinary and aquaculture medicine. It is often used on dairy cattle and aquacultured catfish, salmon, and lobster.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 64-72-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H23 ClN2O8 · HCl
MDL Number MFCD00082440
Melting Point 210°C to 215°C
Synonym Chlortetracycline hydrochloride; 7-Chlorotetracycline hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 515.34
Grade Certified Reference Material