missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002383
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002383 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002383 Supplier Merck Emd Millipore Supplier No. PHR1647200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 1202-34-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H9N3
MDL Number MFCD00006247
Melting Point 90°C to 92°C
Boiling Point 222°C (50 mmHg)
Synonym 2,2′-Dipyridylamine; 2,2′-DIPYRIDYLAMINE
Type Certified Reference Material
Quantity 200 mg
Formula Weight 171.20
Grade Certified Reference Material
Show More Show Less