missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine Impurity A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018259
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018259 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018259 Supplier Merck Emd Millipore Supplier No. PHR200450MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Pharmaceutical (small molecule)

Specifications

CAS 1246816-57-8
Linear Formula C16H24 ClN3
Synonym 2-(4-Chlorophenyl)-4-(dimethylamino)-2-[2-(dimethylamino)ethyl]butanenitrile; 2-(4-CHLOROPHENYL)-4-(DIMETHYLAMINO)-2-[2-(DIMETHYLAMINO)ETHYL]BUTANENITRILE
Type Certified Reference Material
Quantity 50 mg
Formula Weight 293.83
Grade Certified Reference Material