Learn More
Chlorotris(triphenylphosphine)cobalt(I), 97%
CAS: 26305-75-9 | C54H45ClCoP3 | 881.26 g/mol
$92.15 - $317.75
Chemical Identifiers
| CAS | 26305-75-9 |
|---|---|
| Molecular Formula | C54H45ClCoP3 |
| Molecular Weight (g/mol) | 881.26 |
| MDL Number | MFCD00015864 |
| InChI Key | RHNBOEDTKLKUEB-UHFFFAOYSA-M |
| Synonym | chlorotris triphenylphosphine cobalt i, cobalt i tris triphenylphosphine chloride |
| PubChem CID | 71431130 |
| IUPAC Name | cobalt;triphenylphosphane;chloride |
| SMILES | [Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5539503
|
Thermo Scientific Chemicals
H5539503 |
1 g |
Each for $92.15
|
|
|||||
|
AAH5539506
|
Thermo Scientific Chemicals
H5539506 |
5 g |
Each for $317.75
|
|
|||||
Description
A stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA stoichiometric reducing agent that is used in the radical dimerization of halogenated organic molecules.
Solubility
Insoluble in water.
Notes
Air & Moisture Sensitive.
Chemical Identifiers
| 26305-75-9 | |
| 881.26 | |
| RHNBOEDTKLKUEB-UHFFFAOYSA-M | |
| 71431130 | |
| [Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C54H45ClCoP3 | |
| MFCD00015864 | |
| chlorotris triphenylphosphine cobalt i, cobalt i tris triphenylphosphine chloride | |
| cobalt;triphenylphosphane;chloride |
Specifications
| 26305-75-9 | |
| C54H45ClCoP3 | |
| 1 g | |
| chlorotris triphenylphosphine cobalt i, cobalt i tris triphenylphosphine chloride | |
| RHNBOEDTKLKUEB-UHFFFAOYSA-M | |
| cobalt;triphenylphosphane;chloride | |
| 71431130 | |
| 97% |
| 135°C to 139°C | |
| MFCD00015864 | |
| Air and moisture sensitive | |
| Insoluble in water. | |
| [Cl-].[Co+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 881.26 | |
| 881.26 | |
| Chlorotris(triphenylphosphine)cobalt(I) |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only