missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotrimethylsilane 98.0+%, TCI America™
Supplier: TCI America C0306100ML
Specifications
| Chlorotrimethylsilane | |
| -58°C | |
| C3H9ClSi | |
| 100 mL | |
| trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl | |
| C[Si](C)(C)Cl | |
| 108.64 | |
| CHEBI:85069 | |
| ≥98.0% (GC) |
| 75-77-4 | |
| 57°C | |
| MFCD00000502 | |
| 1298 | |
| IJOOHPMOJXWVHK-UHFFFAOYSA-N | |
| chloro(trimethyl)silane | |
| 6397 | |
| 108.64 | |
| Liquid |
Chemical Identifiers
| 75-77-4 | |
| 108.64 | |
| IJOOHPMOJXWVHK-UHFFFAOYSA-N | |
| 6397 | |
| chloro(trimethyl)silane |
| C3H9ClSi | |
| MFCD00000502 | |
| trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl | |
| CHEBI:85069 | |
| C[Si](C)(C)Cl |
Safety and Handling
EINECSNumber : (2)-2041
RTECSNumber : VV2710000
TSCA : Yes