missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane 98.0+%, TCI America™

Catalog No. C030625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C030625ML 25 mL
C0306100ML 100 mL
C0306500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C030625ML Supplier TCI America Supplier No. C030625ML

Chemical Identifiers

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
MDL Number MFCD00000502
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Specifications

Melting Point -58°C
Boiling Point 57°C
Quantity 25 mL
UN Number 1298
Formula Weight 108.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorotrimethylsilane
EINECSNumber (2)-2041
RTECSNumber VV2710000
TSCA Yes