missing translation for 'onlineSavingsMsg'
Learn More

Chlorotrimethylsilane, 98%

CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol

Supplier:  Thermo Scientific Chemicals 110121000

 View more versions of this product

Catalog No. AC110121000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
Specifications

Specifications

Chlorotrimethylsilane
-58.0°C
0.8500g/mL
−28°C
97.5% min. (GC)
C3H9ClSi
(CH3)3SiCl
100 mL
01,1232; 02,435; 03,310; 04,537; 05,709; 06,626; 07,66; 08,107; 09,112; 10,96; 11,127; 12,279; 13,165; 14,88; 15,14; 16,23; 17,79
15,218
Solubility in water: reacts. Other solubilities: care must be taken with ketones,soluble in benzene,ether and perchloroethylene
C[Si](C)(C)Cl
108.64
0.34 mPa.s (25°C)
108.64
Liquid
75-77-4
Colorless
57.0°C
Authentic
Glass bottle
1.3870 to 1.3890
MFCD00000502
04,IV,4007
0.85
trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
IJOOHPMOJXWVHK-UHFFFAOYSA-N
chloro(trimethyl)silane
6397
CHEBI:85069
98%

Chemical Identifiers

75-77-4
108.64
IJOOHPMOJXWVHK-UHFFFAOYSA-N
6397
chloro(trimethyl)silane
C3H9ClSi
MFCD00000502
trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
CHEBI:85069
C[Si](C)(C)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the res

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 200-900-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only