Learn More
Chlorotrimethylsilane, 98%
CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol
Supplier: Thermo Scientific Chemicals 110120010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentSpecifications
| Chlorotrimethylsilane | |
| -58.0°C | |
| 0.8500g/mL | |
| −28°C | |
| 97.5% min. (GC) | |
| C3H9ClSi | |
| (CH3)3SiCl | |
| 1 L | |
| 01,1232; 02,435; 03,310; 04,537; 05,709; 06,626; 07,66; 08,107; 09,112; 10,96; 11,127; 12,279; 13,165; 14,88; 15,14; 16,23; 17,79 | |
| 15,218 | |
| Solubility in water: reacts. Other solubilities: care must be taken with ketones,soluble in benzene,ether and perchloroethylene | |
| C[Si](C)(C)Cl | |
| 108.64 | |
| 0.34 mPa.s (25°C) | |
| 108.64 | |
| Liquid |
| 75-77-4 | |
| Colorless | |
| 57.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3870 to 1.3890 | |
| MFCD00000502 | |
| 04,IV,4007 | |
| 0.85 | |
| trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl | |
| IJOOHPMOJXWVHK-UHFFFAOYSA-N | |
| chloro(trimethyl)silane | |
| 6397 | |
| CHEBI:85069 | |
| 98% |
Chemical Identifiers
| 75-77-4 | |
| 108.64 | |
| IJOOHPMOJXWVHK-UHFFFAOYSA-N | |
| 6397 | |
| chloro(trimethyl)silane |
| C3H9ClSi | |
| MFCD00000502 | |
| trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl | |
| CHEBI:85069 | |
| C[Si](C)(C)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the res
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 200-900-5
RUO – Research Use Only