missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriisopropylsilane, 97+%

CAS: 13154-24-0 | C9H21ClSi | 192.80 g/mol

Supplier:  Thermo Scientific Chemicals A1737618

 View more versions of this product

Catalog No. AAA1737618


Only null left
Explore available promotions

Description

Description

Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Specifications

Specifications

200°C
13154-24-0
C9H21ClSi
UN2987
62°C (144°F)
Miscible with water.
CC(C)[Si](Cl)(C(C)C)C(C)C
192.80
192.81
Acrid
Moisture sensitive
Chlorotriisopropylsilane
50 g
MFCD00009656
1737446
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
KQIADDMXRMTWHZ-UHFFFAOYSA-N
chloro-tri(propan-2-yl)silane
139400
≥97%
1.4535
0.908

Chemical Identifiers

13154-24-0
192.80
KQIADDMXRMTWHZ-UHFFFAOYSA-N
139400
CC(C)[Si](Cl)(C(C)C)C(C)C
C9H21ClSi
MFCD00009656
triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
chloro-tri(propan-2-yl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only