Learn More
Chlorotriisopropylsilane, 97+%
CAS: 13154-24-0 | C9H21ClSi | 192.80 g/mol
Supplier: Thermo Scientific Chemicals A1737609
Description
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Specifications
| 200°C | |
| 13154-24-0 | |
| C9H21ClSi | |
| UN2987 | |
| 62°C (144°F) | |
| Miscible with water. | |
| CC(C)[Si](Cl)(C(C)C)C(C)C | |
| 192.80 | |
| 192.81 | |
| Acrid | |
| Moisture sensitive |
| Chlorotriisopropylsilane | |
| 10 g | |
| MFCD00009656 | |
| 1737446 | |
| triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
| KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
| chloro-tri(propan-2-yl)silane | |
| 139400 | |
| ≥97% | |
| 1.4535 | |
| 0.908 |
Chemical Identifiers
| 13154-24-0 | |
| 192.80 | |
| KQIADDMXRMTWHZ-UHFFFAOYSA-N | |
| 139400 | |
| CC(C)[Si](Cl)(C(C)C)C(C)C |
| C9H21ClSi | |
| MFCD00009656 | |
| triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled | |
| chloro-tri(propan-2-yl)silane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only