missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriethoxysilane 95.0+%, TCI America™
$108.11 - $309.43
Chemical Identifiers
| CAS | 4667-99-6 |
|---|---|
| Molecular Formula | C6H15ClO3Si |
| Molecular Weight (g/mol) | 198.718 |
| MDL Number | MFCD00015124 |
| InChI Key | JEZFASCUIZYYEV-UHFFFAOYSA-N |
| PubChem CID | 78398 |
| IUPAC Name | chloro(triethoxy)silane |
| SMILES | CCO[Si](OCC)(OCC)Cl |
Chemical Identifiers
| 4667-99-6 | |
| 198.718 | |
| JEZFASCUIZYYEV-UHFFFAOYSA-N | |
| chloro(triethoxy)silane |
| C6H15ClO3Si | |
| MFCD00015124 | |
| 78398 | |
| CCO[Si](OCC)(OCC)Cl |
Specifications
| 4667-99-6 | |
| 157°C | |
| MFCD00015124 | |
| 2986 | |
| CCO[Si](OCC)(OCC)Cl | |
| 198.718 | |
| 198.72 | |
| Liquid |
| Colorless | |
| C6H15ClO3Si | |
| 5 g | |
| JEZFASCUIZYYEV-UHFFFAOYSA-N | |
| chloro(triethoxy)silane | |
| 78398 | |
| ≥95.0% (GC) | |
| Chlorotriethoxysilane |
Safety and Handling
EINECSNumber : (2)-2043
TSCA : Yes
Recommended Storage : Refrigerator