missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriethoxysilane 95.0+%, TCI America™
Supplier: TCI America C335425G
Specifications
| Chlorotriethoxysilane | |
| Colorless | |
| C6H15ClO3Si | |
| 25 g | |
| JEZFASCUIZYYEV-UHFFFAOYSA-N | |
| chloro(triethoxy)silane | |
| 78398 | |
| ≥95.0% (GC) |
| 4667-99-6 | |
| 157°C | |
| MFCD00015124 | |
| 2986 | |
| CCO[Si](OCC)(OCC)Cl | |
| 198.718 | |
| 198.72 | |
| Liquid |
Chemical Identifiers
| 4667-99-6 | |
| 198.718 | |
| JEZFASCUIZYYEV-UHFFFAOYSA-N | |
| chloro(triethoxy)silane |
| C6H15ClO3Si | |
| MFCD00015124 | |
| 78398 | |
| CCO[Si](OCC)(OCC)Cl |
Safety and Handling
EINECSNumber : (2)-2043
TSCA : Yes
Recommended Storage : Refrigerator