missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotetracycline, MP Biomedicals™
Chemical Identifiers
| CAS | 64-72-2 |
|---|---|
| Molecular Formula | C22H24Cl2N2O8 |
| Molecular Weight (g/mol) | 515.34 |
| InChI Key | QYAPHLRPFNSDNH-CIVPRPTRSA-N |
| Synonym | Aureomycin, Chlorotetracycline hydrochloride |
| PubChem CID | 66577600 |
| IUPAC Name | (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| SMILES | CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19032705
|
N/A | 5 g |
N/A
|
|
|||||
|
ICN19032780
|
N/A | 100 g |
N/A
|
|
|||||
Chemical Identifiers
| 64-72-2 | |
| 515.34 | |
| Aureomycin, Chlorotetracycline hydrochloride | |
| (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
| C22H24Cl2N2O8 | |
| QYAPHLRPFNSDNH-CIVPRPTRSA-N | |
| 66577600 | |
| CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl |
Specifications
| Chlorotetracycline | |
| 64-72-2 | |
| C22H24Cl2N2O8 | |
| QYAPHLRPFNSDNH-CIVPRPTRSA-N | |
| (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
| 66577600 | |
| Chlorotetracycline |
| Powder | |
| Yellow | |
| Aureomycin, Chlorotetracycline hydrochloride | |
| CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl | |
| 515.34 | |
| 515.3 |