missing translation for 'onlineSavingsMsg'
Learn More

Chlorotetracycline, MP Biomedicals™

Catalog No. ICN19032705
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19032705 5 g
ICN19032780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19032705 Supplier MP Biomedicals Inc Supplier No. 0219032705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-72-2 | Formula: C22H24Cl2N2O8 | Weight: 515.34 g/mol

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym Aureomycin, Chlorotetracycline hydrochloride
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Type Chlorotetracycline
Physical Form Powder
Color Yellow
Quantity 5 g
Formula Weight 515.3
Grade Not Available
Chemical Name or Material Chlorotetracycline