missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorophyll a, ≥95% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 479-61-8 |
| Linear Formula | C55H72 mgN4O5 |
| MDL Number | MFCD00079050 |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 893.49 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |