missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorophenol Red 93.0+%, TCI America™
$31.25 - $40.44
Chemical Identifiers
| CAS | 4430-20-0 |
|---|---|
| Molecular Formula | C19H12Cl2O5S |
| Molecular Weight (g/mol) | 423.26 |
| MDL Number | MFCD00005877 |
| InChI Key | WWAABJGNHFGXSJ-UHFFFAOYSA-N |
| PubChem CID | 20486 |
| IUPAC Name | 3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione |
| SMILES | OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1 |
Chemical Identifiers
| 4430-20-0 | |
| 423.26 | |
| WWAABJGNHFGXSJ-UHFFFAOYSA-N | |
| 3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione |
| C19H12Cl2O5S | |
| MFCD00005877 | |
| 20486 | |
| OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1 |
Specifications
| 4430-20-0 | |
| MFCD00005877 | |
| OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1 | |
| 423.26 | |
| 423.26 | |
| Crystal/Powder |
| C19H12Cl2O5S | |
| WWAABJGNHFGXSJ-UHFFFAOYSA-N | |
| 3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione | |
| 20486 | |
| ≥93.0% (HPLC) | |
| Chlorophenol Red |
Safety and Handling
TSCA : Yes