missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red 93.0+%, TCI America™

Catalog No. C02451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02451G 1 g
C02455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02451G Supplier TCI America Supplier No. C02451G

Chemical Identifiers

CAS 4430-20-0
Molecular Formula C19H12Cl2O5S
Molecular Weight (g/mol) 423.26
MDL Number MFCD00005877
InChI Key WWAABJGNHFGXSJ-UHFFFAOYSA-N
PubChem CID 20486
IUPAC Name 3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1

Specifications

Formula Weight 423.26
Percent Purity ≥93.0% (HPLC)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Chlorophenol Red