missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red, 0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red, Certified, LabChem™

Supplier:  LabChem LC130802

 View more versions of this product

Catalog No. LC130802


Only null left

Description

Description

Specifications

Specifications

Chlorophenol Red
123333-64-2 , 7732-18-5
C19H11Cl2O5S
MFCD00151199
Passes Test
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
422.25
445.25
Amber Glass
Red
Liquid
0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red
99.96,0.04
C19H11Cl2O5SNa
Soluble in water
QXTPRQZMDKBTAI-UNOMPAQXSA-M
2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
131846007
Certified
Hydrogen chloride; Carbon monoxide; Carbon dioxide
1 L

Chemical Identifiers

123333-64-2
422.25
QXTPRQZMDKBTAI-UNOMPAQXSA-M
2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
C19H11Cl2O5S
MFCD00151199
131846007
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions