missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red, 0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red, Certified, LabChem™

Catalog No. LC130807
Change view
Click to view available options
Quantity:
125 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC130807 125 mL
LC130802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC130807 Supplier LabChem Supplier No. LC130807

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123333-64-2 , 7732-18-5
Molecular Formula C19H11Cl2O5S
Molecular Weight (g/mol) 422.25
MDL Number MFCD00151199
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
PubChem CID 131846007
IUPAC Name 2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O

Specifications

CAS Max % 99.96, 0.04
Identification Passes Test
Packaging Amber Glass
Linear Formula C19H11Cl2O5SNa
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Color Red
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 445.25
Grade Certified
Chemical Name or Material Chlorophenol Red, 0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature