missing translation for 'onlineSavingsMsg'
Learn More

Chloropentafluorobenzene 98.0+%, TCI America™

Catalog No. P085025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P085025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P085025G Supplier TCI America Supplier No. P085025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 344-07-0
Molecular Formula C6ClF5
Molecular Weight (g/mol) 202.51
MDL Number MFCD00000534
InChI Key KGCDGLXSBHJAHZ-UHFFFAOYSA-N
Synonym chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
PubChem CID 9579
IUPAC Name 1-chloro-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Cl)C(F)=C1F

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
Formula Weight 202.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloropentafluorobenzene
RTECSNumber CZ1225500
TSCA Yes