missing translation for 'onlineSavingsMsg'
Learn More

Chloropentafluorobenzene, 98+%

CAS: 344-07-0 | C6ClF5 | 202.51 g/mol

$68.85 - $224.19

Chemical Identifiers

CAS 344-07-0
Molecular Formula C6ClF5
Molecular Weight (g/mol) 202.51
MDL Number MFCD00000534
InChI Key KGCDGLXSBHJAHZ-UHFFFAOYSA-N
Synonym chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
PubChem CID 9579
IUPAC Name 1-chloro-2,3,4,5,6-pentafluorobenzene
SMILES FC1=C(F)C(F)=C(Cl)C(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1538409
View Documents
Thermo Scientific Chemicals
A1538409
10 g
Each for $68.85
Only null left
 
AAA1538418
View Documents
Thermo Scientific Chemicals
A1538418
50 g
Each for $224.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

344-07-0
202.51
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
9579
FC1=C(F)C(F)=C(Cl)C(F)=C1F
C6ClF5
MFCD00000534
chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
1-chloro-2,3,4,5,6-pentafluorobenzene

Specifications

344-07-0
116°C to 118°C
1.424
10 g
chloropentafluorobenzene, pentafluorochlorobenzene, benzene, chloropentafluoro, pentafluorophenyl chloride, chloroperfluorobenzene, ccris 2394, pentafluorochloro benzene, 1-chloro-2,3,4,5,6-pentafluoro-benzene, benzene, 1-chloro-2,3,4,5,6-pentafluoro, pentafluorophenylchloride
FC1=C(F)C(F)=C(Cl)C(F)=C1F
202.51
202.51
Chloropentafluorobenzene
1.568
C6ClF5
MFCD00000534
1819389
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
1-chloro-2,3,4,5,6-pentafluorobenzene
9579
≥98%
Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P311-P501c

H331

EINECSNumber : 206-450-6

RTECSNumber : CZ1225500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only