missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylsulfonyl Chloride 95.0+%, TCI America™

Catalog No. p-7124686
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09245G 5 g
C092425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09245G Supplier TCI America Supplier No. C09245G
Only null left

Chemical Identifiers

CAS 3518-65-8
Molecular Formula CH2Cl2O2S
Molecular Weight (g/mol) 148.99
MDL Number MFCD00014727
InChI Key KQDDQXNVESLJNO-UHFFFAOYSA-N
Synonym methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
PubChem CID 77054
IUPAC Name chloromethanesulfonyl chloride
SMILES ClCS(Cl)(=O)=O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
UN Number 3265
Formula Weight 148.99
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Chloromethylsulfonyl Chloride
RTECSNumber PB2800000
TSCA No