accessibility menu, dialog, popup

UserName

CAS RN 3518-65-8

IUPAC Name: chloromethanesulfonyl chloride
Synonyms: chloromethanesulfonyl chloride
Molecular Weight (g/mol): 148.99
Molecular Formula: CH2Cl2O2S
InChi Key: KQDDQXNVESLJNO-UHFFFAOYSA-N
SMILES: ClCS(Cl)(=O)=O
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)