missing translation for 'onlineSavingsMsg'
Learn More

Chloromethylstyrene (m- and p- mixture) (stabilized with TBC + ONP + o-Nitrocresol) 88.0+%, TCI America™

Catalog No. p-7124596
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0767500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0767500G Supplier TCI America Supplier No. C0767500G

Chemical Identifiers

CAS 57458-41-0
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.621
MDL Number MFCD00051362
InChI Key SLBOQBILGNEPEB-UHFFFAOYSA-N
Synonym vinylbenzyl chloride, vinylbenzylchloride, vinyl benzyl chloride, 1-chloroprop-2-en-1-yl benzene, 1-phenylallyl-chloride, 1-chloroallyl benzene, acmc-1b1l3, ksc269g1b, 1-chloranylprop-2-enylbenzene, 3-phenyl-3-chloro-1-propene
PubChem CID 11768772
IUPAC Name 1-chloroprop-2-enylbenzene
SMILES C=CC(C1=CC=CC=C1)Cl

Specifications

Color Yellow
Quantity 500 g
UN Number 3265
Formula Weight 152.62
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethylstyrene (m- and p- mixture) (stabilized with TBC + ONP + o-Nitrocresol)
EINECSNumber (3)-0046
TSCA No
Recommended Storage Refrigerator