accessibility menu, dialog, popup

UserName

CH2Cl

CAS RN 57458-41-0

IUPAC Name: (1-chloroprop-2-en-1-yl)benzene
Synonyms: (1-chloroprop-2-en-1-yl)benzene
Molecular Weight (g/mol): 152.62
Molecular Formula: C9H9Cl
InChi Key: SLBOQBILGNEPEB-UHFFFAOYNA-N
SMILES: ClC(C=C)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results