missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trimethylsilane 98.0+%, TCI America™

Catalog No. p-7124651
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C086225ML 25 mL
C0862100ML 100 mL
C0862250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C086225ML Supplier TCI America Supplier No. C086225ML
Only null left

Chemical Identifiers

CAS 2344-80-1
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00000878
InChI Key OOCUOKHIVGWCTJ-UHFFFAOYSA-N
Synonym chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
PubChem CID 75361
IUPAC Name chloromethyl(trimethyl)silane
SMILES C[Si](C)(C)CCl

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 mL
UN Number 2924
Formula Weight 122.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)trimethylsilane
EINECSNumber (2)-3846
TSCA Yes