accessibility menu, dialog, popup

UserName

CH3SiCH3H3CCl

CAS RN 2344-80-1

IUPAC Name: (chloromethyl)trimethylsilane
Synonyms: (chloromethyl)trimethylsilane
Molecular Weight (g/mol): 122.67
Molecular Formula: C4H11ClSi
InChi Key: OOCUOKHIVGWCTJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCl
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)