missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trimethylsilane, 98%

Catalog No. AAA1430330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1430330 250 g
AAA1430309 10 g
AAA1430318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1430330 Supplier Thermo Scientific Chemicals Supplier No. A1430330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2344-80-1 | C4H11ClSi | 122.667 g/mol

(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

Solubility
Soluble in water (0.4g/L at 20°C).

Notes
Store in cool, well ventilated area. Keep container tightly closed. Moisture sensitive. Store under argon.

Chemical Identifiers

CAS 2344-80-1
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00000878
InChI Key OOCUOKHIVGWCTJ-UHFFFAOYSA-N
Synonym chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
PubChem CID 75361
IUPAC Name chloromethyl(trimethyl)silane
SMILES C[Si](C)(C)CCl

Specifications

Boiling Point 97°C to 98°C
Quantity 250 g
UN Number UN1993
Beilstein 906705
Flash Point −2°C (28°F)
Solubility Information Soluble in water (0.4g/L at 20°C).
Formula Weight 122.67
Percent Purity 98%
Odor Odorless
Refractive Index 1.418
Sensitivity Moisture sensitive
Density 0.886
Chemical Name or Material (Chloromethyl)trimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-058-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only