missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trimethoxysilane 98.0+%, TCI America™

Catalog No. C307325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C307325G 25 g
C30735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C307325G Supplier TCI America Supplier No. C307325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5926-26-1
Molecular Formula C4H11ClO3Si
Molecular Weight (g/mol) 170.66
MDL Number MFCD00468107
InChI Key FPOSCXQHGOVVPD-UHFFFAOYSA-N
PubChem CID 22212
IUPAC Name (chloromethyl)trimethoxysilane
SMILES CO[Si](CCl)(OC)OC

Specifications

Color Colorless
Boiling Point 156°C
Quantity 25 g
UN Number 1993
Formula Weight 170.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)trimethoxysilane
EINECSNumber (2)-2079
RTECSNumber VV2625000
TSCA No